missing translation for 'onlineSavingsMsg'
Learn More

Phenylacetic Hydrazide 98.0+%, TCI America™

Catalog No. P22035G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P22035G 5 g
P220325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P22035G Supplier TCI America Supplier No. P22035G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 937-39-3
Molecular Formula C8H10N2O
Molecular Weight (g/mol) 150.181
MDL Number MFCD00007612
InChI Key FPTCVTJCJMVIDV-UHFFFAOYSA-N
Synonym phenylacetic acid hydrazide, benzeneacetic acid, hydrazide, phenylacetic hydrazide, phenylacetylhydrazine, phenylacetyl hydrazide, phenacetic acid hydrazide, 2-phenylacetyl hydrazine, phenylacetohydrazide, phenylacethydrazide, phenyl-acetic acid hydrazide
PubChem CID 70301
IUPAC Name 2-phenylacetohydrazide
SMILES C1=CC=C(C=C1)CC(=O)NN

Specifications

Melting Point 117°C
Quantity 5 g
Formula Weight 150.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Phenylacetic Hydrazide