missing translation for 'onlineSavingsMsg'
Learn More

Phenylacetaldehyde (40-55% in Diethyl Phthalate), TCI America™

Catalog No. P0119100G
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0119100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P0119100G Supplier TCI America Supplier No. P0119100G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 122-78-1
Molecular Formula C8H8O
Molecular Weight (g/mol) 120.15
MDL Number MFCD00006993
InChI Key DTUQWGWMVIHBKE-UHFFFAOYSA-N
Synonym phenylacetaldehyde, benzeneacetaldehyde, hyacinthin, alpha-tolualdehyde, phenylethanal, 2-phenylethanal, phenylacetic aldehyde, alpha-toluic aldehyde, acetaldehyde, phenyl, benzylcarboxaldehyde
PubChem CID 998
ChEBI CHEBI:16424
IUPAC Name 2-phenylacetaldehyde
SMILES O=CCC1=CC=CC=C1

Specifications

Color Yellow
Quantity 100 g
Formula Weight 120.15
Physical Form Liquid
Chemical Name or Material Phenylacetaldehyde (40-55% in Diethyl Phthalate)
EINECSNumber (3)-2656
RTECSNumber CY1420000
TSCA Yes
Recommended Storage Refrigerator