missing translation for 'onlineSavingsMsg'
Learn More

Phenyl Trifluoromethyl Sulfone 98.0+%, TCI America™

Catalog No. P21955G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P21955G 5 g
P21951G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P21955G Supplier TCI America Supplier No. P21955G

Chemical Identifiers

CAS 426-58-4
Molecular Formula C7H5F3O2S
Molecular Weight (g/mol) 210.17
MDL Number MFCD00159083
InChI Key UPGBQYFXKAKWQC-UHFFFAOYSA-N
PubChem CID 555605
IUPAC Name trifluoromethylsulfonylbenzene
SMILES C1=CC=C(C=C1)S(=O)(=O)C(F)(F)F

Specifications

Color Colorless
Boiling Point 203°C
Quantity 5 g
Formula Weight 210.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Phenyl Trifluoromethyl Sulfone