missing translation for 'onlineSavingsMsg'
Learn More

Phenyl Trifluoromethyl Sulfide 97.0+%, TCI America™

Catalog No. P16935G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P16935G 5 g
P169325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P16935G Supplier TCI America Supplier No. P16935G

Chemical Identifiers

CAS 456-56-4
Molecular Formula C7H5F3S
Molecular Weight (g/mol) 178.17
MDL Number MFCD00040839
InChI Key YQQKTCBMKQQOSM-UHFFFAOYSA-N
Synonym phenyl trifluoromethyl sulfide, trifluoromethylthiobenzene, trifluoromethylthio benzene, trifluoromethyl thio benzene, benzene, trifluoromethyl thio, trifluoromethyl phenyl sulfide, phenyl trifluoromethyl sulphide, pubchem10400, phenyl trifluoromethylsulfide
PubChem CID 68011
IUPAC Name [(trifluoromethyl)sulfanyl]benzene
SMILES FC(F)(F)SC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 142°C
Quantity 5 g
UN Number 1993
Formula Weight 178.17
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Phenyl Trifluoromethyl Sulfide