missing translation for 'onlineSavingsMsg'
Learn More

Phenyl trifluoromethyl sulfide, 98%

Catalog No. AAL1011406
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1011406 5 g
AAL1011403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1011406 Supplier Thermo Scientific Chemicals Supplier No. L1011406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 456-56-4 | C7H5F3S | 178.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 456-56-4
Molecular Formula C7H5F3S
Molecular Weight (g/mol) 178.17
MDL Number MFCD00040839
InChI Key YQQKTCBMKQQOSM-UHFFFAOYSA-N
Synonym phenyl trifluoromethyl sulfide, trifluoromethylthiobenzene, trifluoromethylthio benzene, trifluoromethyl thio benzene, benzene, trifluoromethyl thio, trifluoromethyl phenyl sulfide, phenyl trifluoromethyl sulphide, pubchem10400, phenyl trifluoromethylsulfide
PubChem CID 68011
IUPAC Name trifluoromethylsulfanylbenzene
SMILES FC(F)(F)SC1=CC=CC=C1

Specifications

Density 1.249
Boiling Point 140°C to 142°C
Flash Point 37°C (98°F)
Odor Unpleasant
Refractive Index 1.4655
Quantity 5 g
UN Number UN1993
Beilstein 2043134
Formula Weight 178.18
Percent Purity 98%
Chemical Name or Material Phenyl trifluoromethyl sulfide
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 207-270-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only