missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenyl Sulfide 98.0+%, TCI America™
Supplier: TCI America P023025G
Specifications
| Phenyl Sulfide | |
| -40°C | |
| 296°C | |
| MFCD00003064 | |
| 3082 | |
| LTYMSROWYAPPGB-UHFFFAOYSA-N | |
| (phenylsulfanyl)benzene | |
| 8766 | |
| 186.27 | |
| Liquid |
| 139-66-2 | |
| Yellow | |
| C12H10S | |
| 25 g | |
| diphenyl sulfide, phenyl sulfide, phenylthiobenzene, diphenyl sulphide, diphenyl thioether, benzene, 1,1'-thiobis, diphenylmercaptan, diphenylthiamethane, diphenyl monosulfide, sulfide, diphenyl | |
| S(C1=CC=CC=C1)C1=CC=CC=C1 | |
| 186.27 | |
| CHEBI:38959 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 139-66-2 | |
| 186.27 | |
| LTYMSROWYAPPGB-UHFFFAOYSA-N | |
| 8766 | |
| (phenylsulfanyl)benzene |
| C12H10S | |
| MFCD00003064 | |
| diphenyl sulfide, phenyl sulfide, phenylthiobenzene, diphenyl sulphide, diphenyl thioether, benzene, 1,1'-thiobis, diphenylmercaptan, diphenylthiamethane, diphenyl monosulfide, sulfide, diphenyl | |
| CHEBI:38959 | |
| S(C1=CC=CC=C1)C1=CC=CC=C1 |
Safety and Handling
EINECSNumber : (3)-1109
RTECSNumber : SX2275000
TSCA : Yes