missing translation for 'onlineSavingsMsg'
Learn More

Phenyl Sulfide 98.0+%, TCI America™

Catalog No. p-7134503
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P023025G 25 g
P0230500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P023025G Supplier TCI America Supplier No. P023025G
Only null left

Chemical Identifiers

CAS 139-66-2
Molecular Formula C12H10S
Molecular Weight (g/mol) 186.27
MDL Number MFCD00003064
InChI Key LTYMSROWYAPPGB-UHFFFAOYSA-N
Synonym diphenyl sulfide, phenyl sulfide, phenylthiobenzene, diphenyl sulphide, diphenyl thioether, benzene, 1,1'-thiobis, diphenylmercaptan, diphenylthiamethane, diphenyl monosulfide, sulfide, diphenyl
PubChem CID 8766
ChEBI CHEBI:38959
IUPAC Name (phenylsulfanyl)benzene
SMILES S(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point -40°C
Color Yellow
Boiling Point 296°C
Quantity 25 g
UN Number 3082
Formula Weight 186.27
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Phenyl Sulfide
EINECSNumber (3)-1109
RTECSNumber SX2275000
TSCA Yes