accessibility menu, dialog, popup

UserName

S

CAS RN 139-66-2

IUPAC Name: (phenylsulfanyl)benzene
Synonyms: phenyl sulfide|1,1'-sulfanediyldibenzene
Molecular Weight (g/mol): 186.27
Molecular Formula: C12H10S
InChi Key: LTYMSROWYAPPGB-UHFFFAOYSA-N
SMILES: S(C1=CC=CC=C1)C1=CC=CC=C1
Boiling Point 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (4)