missing translation for 'onlineSavingsMsg'
Learn More

Phenyl salicylate melting point Standard Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111012696
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012696 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012696 Supplier Merck Emd Millipore Supplier No. PHR11521G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.Phenyl salicylate is a phenyl ester of salicylic acid and is used as a medicine under the name ′salol′ as an internal antiseptic. It finds its application in paints, waxes, and varnishes as it is found to absorb UV radiation in the range of 290-325 nm. It also shows its presence in cosmetics and sunscreens as a UV filter.

Specifications

CAS 118-55-8
Flash Point 113°C - closed cup
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula 2-(HO)C6H4 CO2C6H5
MDL Number MFCD00002213
Melting Point ∼42°C; 41°C to 43°C
Boiling Point 172°C to 173°C (12 mmHg)
Synonym Phenyl salicylate; Salol
Type Certified Reference Material
Quantity 1 g
Formula Weight 214.22
Grade Certified Reference Material
Show More Show Less