missing translation for 'onlineSavingsMsg'
Learn More

Phenyl Salicylate, Crystal, Purified, 98%, Spectrum™ Chemical

Catalog No. 18612288
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
40 kg
50 kg
Packaging:
Poly Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18612288 50 kg Poly Bottle
18612283 125 g Poly Bottle
18612284 500 g Poly Bottle
18612285 2.5 kg Poly Bottle
18612286 12 kg Poly Bottle
18612287 40 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18612288 Supplier Spectrum Chemical Mfg Cor Supplier No. PH16050KGBL
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 118-55-8

Specifications

CAS Max % 1
Melting Point 40° to 47°C
Boiling Point 172°C to 173°C/341.6°F to 343.4°F
Packaging Poly Bottle
Quantity 50 kg
Molecular Weight (g/mol) 214.22
Percent Purity 0.98
Grade Purified
Physical Form Crystals or Flakes
Chemical Name or Material Phenyl Salicylate, Crystal

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.