missing translation for 'onlineSavingsMsg'
Learn More

Phenyl p-Toluenesulfonate 99.0+%, TCI America™

Catalog No. P023925G
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P023925G 25 g
P023910G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P023925G Supplier TCI America Supplier No. P023925G

Chemical Identifiers

CAS 640-60-8
Molecular Formula C13H12O3S
Molecular Weight (g/mol) 248.296
MDL Number MFCD00025996
InChI Key KZQFPRKQBWRRHQ-UHFFFAOYSA-N
Synonym phenyl para-toluenesulfonate, p-toluenesulfonic acid phenyl ester, phenyl tosylate, phenyl p-toluenesulfonate, phenylp-toluenesulfonate, acmc-209njz, phenyl 4-methylbenzene-1-sulfonate, toluene-4-sulfonic acid phenyl ester, benzenesulfonic acid,4-methyl-, phenyl ester
PubChem CID 223146
IUPAC Name phenyl 4-methylbenzenesulfonate
SMILES CC1=CC=C(C=C1)S(=O)(=O)OC2=CC=CC=C2

Specifications

Melting Point 96°C
Color White
Quantity 25 g
Formula Weight 248.30
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Phenyl p-Toluenesulfonate
EINECSNumber (3)-4062
TSCA No