missing translation for 'onlineSavingsMsg'
Learn More

Phenyl isocyanate, ≥99.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019027
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019027 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019027 Supplier Merck Emd Millipore Supplier No. 78750100ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Phenyl isocyanate is an aromatic monoisocyanate which is a constituent of diphenyl methane diisocyanate. It is suitable to make aryl ureas with amine at room temperature making it unlikely to detect isocyanate formation using NMR. It reacts with amino acids in alkaline solution and with peptone.

Chemical Identifiers

MDL Number MFCD00001994

Specifications

Melting Point -30°C
Density 1.096 g/mL (at 25°C)
CAS 103-71-9
Boiling Point 162°C to 163°C
Flash Point 51°C
Linear Formula C6H5 NCO
Quantity 100 mL
Formula Weight 119.12
Vapor Pressure 1.4 mmHg (20°C)
Percent Purity ≥99.0% (GC)
Grade For HPLC derivatization
Show More Show Less
RTECSNumber DA3675000