missing translation for 'onlineSavingsMsg'
Learn More

Phenyl Chloroformate 98.0+%, TCI America™

Catalog No. C0649500ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0649500ML 500 mL
C064925ML 25 mL
C0649100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C0649500ML Supplier TCI America Supplier No. C0649500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1885-14-9
Molecular Formula C7H5ClO2
Molecular Weight (g/mol) 156.57
MDL Number MFCD00000637
InChI Key AHWALFGBDFAJAI-UHFFFAOYSA-N
Synonym phenyl chloroformate, chloroformic acid phenyl ester, phenyl chlorocarbonate, carbonochloridic acid, phenyl ester, phenoxycarbonyl chloride, formic acid, chloro-, phenyl ester, fenylester kyseliny chlormravenci, phenylchloroformate, unii-6tnd0d6d3y, 6tnd0d6d3y
PubChem CID 15891
IUPAC Name phenyl carbonochloridate
SMILES ClC(=O)OC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 187°C
Quantity 500 mL
UN Number 2746
Formula Weight 156.57
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Phenyl Chloroformate
EINECSNumber (3)-0629
RTECSNumber FG3850000
TSCA Yes
Recommended Storage Refrigerator