missing translation for 'onlineSavingsMsg'
Learn More

Phenyl chloroformate, 99%

Catalog No. AC218212500
Change view
Click to view available options
Quantity:
250 mL
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC218212500 250 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC218212500 Supplier Thermo Scientific Chemicals Supplier No. 218212500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1885-14-9 | C7H5ClO2 | 156.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent

Chemical Identifiers

CAS 1885-14-9
Molecular Formula C7H5ClO2
Molecular Weight (g/mol) 156.57
MDL Number MFCD00000637
InChI Key AHWALFGBDFAJAI-UHFFFAOYSA-N
Synonym phenyl chloroformate, chloroformic acid phenyl ester, phenyl chlorocarbonate, carbonochloridic acid, phenyl ester, phenoxycarbonyl chloride, formic acid, chloro-, phenyl ester, fenylester kyseliny chlormravenci, phenylchloroformate, unii-6tnd0d6d3y, 6tnd0d6d3y
PubChem CID 15891
IUPAC Name phenyl carbonochloridate
SMILES ClC(=O)OC1=CC=CC=C1

Specifications

Melting Point -28.0°C
Density 1.2440g/mL
Boiling Point 188.0°C to 189.0°C (1013.0 mbar)
Flash Point 80°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5100 to 1.5120
Linear Formula ClCO2C6H5
Quantity 250 mL
Specific Gravity 1.244
Solubility Information Solubility in water: hydrolysis
Viscosity 1.86 mPa.s (20°C)
Formula Weight 156.57
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Phenyl chloroformate
Show More Show Less
Hazard Category GHS Signal Word: Danger
EINECSNumber 217-547-8

RUO – Research Use Only