missing translation for 'onlineSavingsMsg'
Learn More

Phenyl beta-D-Glucopyranoside Hydrate 99.0+%, TCI America™

Catalog No. P01781G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P01781G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P01781G Supplier TCI America Supplier No. P01781G

Chemical Identifiers

CAS 1464-44-4
Molecular Formula C12H16O6
Molecular Weight (g/mol) 256.25
MDL Number MFCD03410292
InChI Key NEZJDVYDSZTRFS-WLEIBRHLNA-N
Synonym phenyl beta-d-glucopyranoside, phenylglucoside, phenyl-beta-d-glucopyranoside, phenol glucoside, phenyl beta-d-glucoside, phenyl-beta-d-glucoside, .beta.-d-glucopyranoside, phenyl, unii-4w3pgi3766, aryl beta-d-glucoside, glucopyranoside, phenyl-, beta-d
PubChem CID 65080
ChEBI CHEBI:28749
IUPAC Name (2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-phenoxyoxane-3,4,5-triol
SMILES OC[C@H]1O[C@@H](OC2=CC=CC=C2)[C@H](O)[C@@H](O)[C@@H]1O

Specifications

Melting Point 173°C
Color White
Quantity 1 g
Formula Weight 256.25
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Phenyl beta-D-Glucopyranoside Hydrate
RTECSNumber LZ5985510
TSCA Yes