missing translation for 'onlineSavingsMsg'
Learn More

Phenyl beta-D-glucopyranoside, 98%

Catalog No. AAJ6688514
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6688514 25 g
AAJ6688506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6688514 Supplier Thermo Scientific Chemicals Supplier No. J6688514

A substrate used for the determination of →-D-glucosidase | CAS: 1464-44-4 | C12H16O6 | 256.25 g/mol

A substrate used for the determination of D-glucosidase

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1464-44-4
Molecular Formula C12H16O6
Molecular Weight (g/mol) 256.25
MDL Number MFCD03410292
InChI Key NEZJDVYDSZTRFS-WLEIBRHLNA-N
Synonym phenyl beta-d-glucopyranoside, phenylglucoside, phenyl-beta-d-glucopyranoside, phenol glucoside, phenyl beta-d-glucoside, phenyl-beta-d-glucoside, .beta.-d-glucopyranoside, phenyl, unii-4w3pgi3766, aryl beta-d-glucoside, glucopyranoside, phenyl-, beta-d
PubChem CID 65080
ChEBI CHEBI:28749
IUPAC Name (2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-phenoxyoxane-3,4,5-triol
SMILES OC[C@H]1O[C@@H](OC2=CC=CC=C2)[C@H](O)[C@@H](O)[C@@H]1O

Specifications

Melting Point 173°C to 176°C
Color White
Quantity 25 g
Beilstein 87517
Solubility Information Soluble in water
Optical Rotation −73° (c=1 in Water)
Formula Weight 256.25
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Phenyl beta-D-glucopyranoside
EINECSNumber 215-978-6
RTECSNumber LZ5985510
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only