Learn More
Phenoxybenzamine hydrochloride
CAS: 63-92-3 | C18H23Cl2NO | 340.288 g/mol
Supplier: Thermo Scientific Chemicals J6190003
Description
Selective alpha adrenergic blocking agent. Irreversible calmodulin inhibitor
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Selective alpha adrenergic blocking agent
- Irreversible calmodulin inhibitor
Specifications
| Phenoxybenzamine hydrochloride | |
| White | |
| MFCD00055152 | |
| 14,7256 | |
| VBCPVIWPDJVHAN-UHFFFAOYSA-N | |
| N-benzyl-N-(2-chloroethyl)-1-phenoxypropan-2-amine;hydrochloride | |
| 5284441 | |
| 340.29 |
| 63-92-3 | |
| C18H23Cl2NO | |
| 1 g | |
| phenoxybenzamine hydrochloride, dibenzyline, phenoxybenzamine hcl, n-benzyl-n-2-chloroethyl-1-phenoxypropan-2-amine hydrochloride, dibenzyline hydrochloride, phenoxybenzamine.hcl, bensylyt nen, phenoxybenzamine chloride, benzene methanamine | |
| CC(COC1=CC=CC=C1)N(CCCl)CC2=CC=CC=C2.Cl | |
| 340.288 | |
| CHEBI:8078 | |
| Powder |
Chemical Identifiers
| 63-92-3 | |
| 340.288 | |
| VBCPVIWPDJVHAN-UHFFFAOYSA-N | |
| 5284441 | |
| N-benzyl-N-(2-chloroethyl)-1-phenoxypropan-2-amine;hydrochloride |
| C18H23Cl2NO | |
| MFCD00055152 | |
| phenoxybenzamine hydrochloride, dibenzyline, phenoxybenzamine hcl, n-benzyl-n-2-chloroethyl-1-phenoxypropan-2-amine hydrochloride, dibenzyline hydrochloride, phenoxybenzamine.hcl, bensylyt nen, phenoxybenzamine chloride, benzene methanamine | |
| CHEBI:8078 | |
| CC(COC1=CC=CC=C1)N(CCCl)CC2=CC=CC=C2.Cl |
Safety and Handling
GHS H Statement
H351-H302
Suspected of causing cancer.
Harmful if swallowed.
P201-P202-P264b-P270-P281-P301+P312-P308+P313-P330-P501c
H302-H350
EINECSNumber : 200-569-7
RTECSNumber : DP3750000
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only