missing translation for 'onlineSavingsMsg'
Learn More

Phenoxybenzamine hydrochloride

Catalog No. p-7042261
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ61900MD 250 mg
AAJ6190003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ61900MD Supplier Thermo Scientific Chemicals Supplier No. J61900MD
Only null left

CAS: 63-92-3 | C18H23Cl2NO | 340.288 g/mol

Selective alpha adrenergic blocking agent. Irreversible calmodulin inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Selective alpha adrenergic blocking agent
  • Irreversible calmodulin inhibitor

Chemical Identifiers

CAS 63-92-3
Molecular Formula C18H23Cl2NO
Molecular Weight (g/mol) 340.288
MDL Number MFCD00055152
InChI Key VBCPVIWPDJVHAN-UHFFFAOYSA-N
Synonym phenoxybenzamine hydrochloride, dibenzyline, phenoxybenzamine hcl, n-benzyl-n-2-chloroethyl-1-phenoxypropan-2-amine hydrochloride, dibenzyline hydrochloride, phenoxybenzamine.hcl, bensylyt nen, phenoxybenzamine chloride, benzene methanamine
PubChem CID 5284441
ChEBI CHEBI:8078
IUPAC Name N-benzyl-N-(2-chloroethyl)-1-phenoxypropan-2-amine;hydrochloride
SMILES CC(COC1=CC=CC=C1)N(CCCl)CC2=CC=CC=C2.Cl

Specifications

Color White
Quantity 250 mg
Merck Index 14,7256
Formula Weight 340.29
Physical Form Powder
Chemical Name or Material Phenoxybenzamine hydrochloride
Hazard Category H302-H350
Hazard Statement GHS H Statement
H351-H302
Suspected of causing cancer.
Harmful if swallowed.
Precautionary Statement P201-P202-P264b-P270-P281-P301+P312-P308+P313-P330-P501c
EINECSNumber 200-569-7
RTECSNumber DP3750000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only