accessibility menu, dialog, popup

UserName

CAS RN 63-92-3

IUPAC Name: hydrogen benzyl(2-chloroethyl)(1-phenoxypropan-2-yl)amine chloride
Synonyms: hydrogen phenoxybenzamine chloride
Molecular Weight (g/mol): 340.29
Molecular Formula: C18H23Cl2NO
InChi Key: VBCPVIWPDJVHAN-UHFFFAOYNA-N
SMILES: [H+].[Cl-].CC(COC1=CC=CC=C1)N(CCCl)CC1=CC=CC=C1
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)