missing translation for 'onlineSavingsMsg'
Learn More

Phenoxyacetonitrile 98.0+%, TCI America™

Catalog No. P22175G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P22175G 5 g
P221725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P22175G Supplier TCI America Supplier No. P22175G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3598-14-9
Molecular Formula C8H7NO
Molecular Weight (g/mol) 133.15
MDL Number MFCD00052017
InChI Key VLLSCJFPVSQXDM-UHFFFAOYSA-N
Synonym phenoxyacetonitrile, acetonitrile, phenoxy, 2-phenoxyethanenitrile, phenoxy-acetonitrile, 2-phenoxy-acetonitrile, acmc-1ajch, acetonitrile,2-phenoxy
PubChem CID 241641
IUPAC Name 2-phenoxyacetonitrile
SMILES N#CCOC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 112°C
Quantity 5 g
Formula Weight 133.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Phenoxyacetonitrile