missing translation for 'onlineSavingsMsg'
Learn More

Phenolsulfonic Acid, 65%, Spectrum™ Chemical

Numéro de catalogue. 18605102
Change view
Click to view available options
Quantity:
500 mL
2.5 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Glass Bottle
Poly Drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity Packaging
18605102 200 L Drum
18605099 500 mL Amber Glass Bottle
18605100 2.5 L Glass Bottle
18605101 20 L Poly Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Numéro de catalogue. 18605102 Fournisseur Spectrum Chemical Mfg Cor Code fournisseur PH136200LTBL

Pour de laide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

PH136, 1333-39-7, HOC6H4SO3H

Spectrum™ Chemical Phenolsulfonic Acid

Identifiants chimiques

CAS 1333-39-7
Molecular Formula C6H6O4S
Molecular Weight (g/mol) 174.17
InChI Key IULJSGIJJZZUMF-UHFFFAOYSA-N
IUPAC Name 2-hydroxybenzene-1-sulfonic acid
SMILES OC1=CC=CC=C1S(O)(=O)=O

Spécifications

CAS Max % 1
Packaging Drum
Quantity 200 L
Percent Purity 65%
Grade Reagent