missing translation for 'onlineSavingsMsg'
Learn More

Phenolphthalein StandAR, Macron Fine Chemicals™

Catalog No. MKH306100
Change view
Click to view available options
Quantity:
100mL
Packaging:
Glass
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MKH306100 100mL Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MKH306100 Supplier Avantor Supplier No. H30601

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 77-09-8
Molecular Formula C20H14O4
Molecular Weight (g/mol) 318.33
MDL Number MFCD00005913
InChI Key KJFMBFZCATUALV-UHFFFAOYSA-N
PubChem CID 4764
ChEBI CHEBI:34914
IUPAC Name 3,3-bis(4-hydroxyphenyl)-1,3-dihydro-2-benzofuran-1-one
SMILES OC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1

Specifications

Assay Percent Range 1.0% in 95% Alcohol (APHA)
Packaging Glass
Quantity 100mL
Chemical Name or Material Phenolphthalein, pH 8.0