missing translation for 'onlineSavingsMsg'
Learn More

Phenolphthalein Indicator, 1% (w/v) in Methanol, Neutralized, Ricca Chemical

$161.63 - $236.50

Chemical Identifiers

CAS 77-09-8
Molecular Formula C20H14O4
Molecular Weight (g/mol) Mixture
MDL Number MFCD00005913
InChI Key KJFMBFZCATUALV-UHFFFAOYSA-N
PubChem CID 4764
ChEBI CHEBI:34914
IUPAC Name 3,3-bis(4-hydroxyphenyl)-1,3-dihydro-2-benzofuran-1-one
SMILES OC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
5621-16
View Documents
Ricca Chemical Company
562116
500 mL Natural Poly Bottle
Each for $161.63
Only null left
 
R56210001A
View Documents
Ricca Chemical Company
R56210001A
1 L Natural Poly Bottle
Each for $236.50
Only null left
 
Description

Description

Specifications

Chemical Identifiers

77-09-8
Mixture
KJFMBFZCATUALV-UHFFFAOYSA-N
CHEBI:34914
OC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1
C20H14O4
MFCD00005913
4764
3,3-bis(4-hydroxyphenyl)-1,3-dihydro-2-benzofuran-1-one

Specifications

77-09-8 , 67-56-1 , 7732-18-5
100.0,1.5
C20H14O4
KJFMBFZCATUALV-UHFFFAOYSA-N
3,3-bis(4-hydroxyphenyl)-1,3-dihydro-2-benzofuran-1-one
4764
1% W/V
Natural Poly Bottle
Colorless to Pink
500 mL
Phenolphthalein Indicator
97.0,0.5
1% (w/v)
MFCD00005913
OC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1
Mixture
CHEBI:34914
Indicator
Methanol
8
Liquid
Videos
SDS
Documents

Documents