missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenolphthalein Indicator, 1% (w/v) in Methanol, Neutralized, Ricca Chemical
Supplier: Ricca Chemical Company R56210001A
Specifications
| Phenolphthalein Indicator | |
| 97.0,0.5 | |
| C20H14O4 | |
| KJFMBFZCATUALV-UHFFFAOYSA-N | |
| 3,3-bis(4-hydroxyphenyl)-1,3-dihydro-2-benzofuran-1-one | |
| 4764 | |
| 1% W/V | |
| Natural Poly Bottle | |
| 8 | |
| Liquid |
| 77-09-8 , 67-56-1 , 7732-18-5 | |
| 100.0,1.5 | |
| MFCD00005913 | |
| OC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1 | |
| Mixture | |
| CHEBI:34914 | |
| Indicator | |
| Colorless to Pink | |
| 1 L |
Chemical Identifiers
| 77-09-8 | |
| Mixture | |
| KJFMBFZCATUALV-UHFFFAOYSA-N | |
| CHEBI:34914 | |
| OC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1 |
| C20H14O4 | |
| MFCD00005913 | |
| 4764 | |
| 3,3-bis(4-hydroxyphenyl)-1,3-dihydro-2-benzofuran-1-one |