missing translation for 'onlineSavingsMsg'
Learn More

Phenolphthalein Indicator, 1% (w/v) in Methanol, Neutralized, Ricca Chemical

Catalog No. 562116
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
562116 500 mL Natural Poly Bottle
R56210001A 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 562116 Supplier Ricca Chemical Company Supplier No. 562116

Chemical Identifiers

CAS 77-09-8 , 67-56-1 , 7732-18-5
Molecular Formula C20H14O4
Molecular Weight (g/mol) Mixture
MDL Number MFCD00005913
InChI Key KJFMBFZCATUALV-UHFFFAOYSA-N
PubChem CID 4764
ChEBI CHEBI:34914
IUPAC Name 3,3-bis(4-hydroxyphenyl)-1,3-dihydro-2-benzofuran-1-one
SMILES OC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1

Specifications

CAS Min % 97.0, 0.5
CAS Max % 100.0, 1.5
Concentration 1% (w/v)
Percent Purity 1% W/V
Grade Indicator
Packaging Natural Poly Bottle
Solvent or Matrix Methanol
Color Colorless to Pink
pH 8
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material Phenolphthalein Indicator
Show More Show Less