missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Phenolphthalein, 98.5%, pure, Indicator grade
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
Specifications
Specifications
| Melting Point | 258.0°C to 263.0°C |
| Infrared Spectrum | Authentic |
| Loss on Drying | 0.5% max. (105°C) |
| Packaging | Plastic bottle |
| Assay Percent Range | 98.5% |
| Beilstein | 18, 143 |
| Density | 1.299 |
| Merck Index | 15, 7356 |
| Color | White |
| Solubility Information | Solubility in water: <0.1%. Other solubilities: 1g/12mL alcohol, 1g/100mL ether, soluble in alkalies, soluble in chloroform |
| Show More |
Safety and Handling
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement: May cause cancer. Suspected of causing genetic defects. Suspected of damaging fertility. |
| Precautionary Statement | GHS P Statement: Obtain special instructions before use. IF exposed or concerned: Get medical advice/attention. Use personal protective equipment as required. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
| EINECSNumber | 201-004-7 |
| RTECSNumber | SM8380000 |
| TSCA | TSCA |
RUO – Research Use Only