missing translation for 'onlineSavingsMsg'
Learn More

Phenol 99.5+%, TCI America™

Catalog No. P161025G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P161025G 25 g
P1610500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P161025G Supplier TCI America Supplier No. P161025G

Chemical Identifiers

CAS 108-95-2
Molecular Formula C6H6O
Molecular Weight (g/mol) 94.11
MDL Number MFCD00002143
InChI Key ISWSIDIOOBJBQZ-UHFFFAOYSA-N
PubChem CID 996
ChEBI CHEBI:15882
IUPAC Name phenol
SMILES OC1=CC=CC=C1

Specifications

Melting Point 42°C
Boiling Point 181°C
UN Number 1671
Formula Weight 94.11
Percent Purity ≥99.5% (GC)
Color Red/White
pH 6.0 (H2O soln.)
Quantity 25 g
Physical Form Crystal/Lump
Chemical Name or Material Phenol
EINECSNumber (3)-0481
RTECSNumber SJ3325000
TSCA Yes