missing translation for 'onlineSavingsMsg'
Learn More

Phenol Standard, Certified, 1000ppm ±10ppm (1mL = 1mg), LabChem™

Phenol Standard, C6H6O, CAS Number-108-95-2, 7732-18-5, 500mL, 99.9, 0.1, CHEBI:15882, Colorless, 1000ppm +/-10ppm (1mL - 1mg), 1g/mL, 94.11, Certified, GHS H Statement Solution is not hazardous.

$70.25 - $101.46

Specifications

Chemical Name or Material Phenol Standard
Name Note 1000ppm (1mL = 1mg)
Packaging Amber Glass
CAS 108-95-2 , 7732-18-5
CAS Max % 99.9,0.1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
LC183307
View Documents
LabChem
LC183307
125 mL
Each for $70.25
Only null left
 
LC183301
View Documents
LabChem
LC183301
500 mL
Each for $101.46
Only null left
 
Description

Description

Phenol Standard, C6H6O, CAS Number-108-95-2, 7732-18-5, 500mL, 99.9, 0.1, CHEBI:15882, Colorless, 1000ppm +/-10ppm (1mL - 1mg), 1g/mL, 94.11, Certified, GHS H Statement

Solution is not hazardous.

Specifications

Specifications

Phenol Standard
Amber Glass
99.9,0.1
1g/mL
C6H6O
C6H5O8
Colorless
ISWSIDIOOBJBQZ-UHFFFAOYSA-N
phenol
996
94.11
1000ppm (1mL = 1mg)
108-95-2 , 7732-18-5
Passes Test
Liquid
1000 ppm ±10 ppm (1mL = 1mg)
MFCD00002143
Soluble in water
OC1=CC=CC=C1
94.11
CHEBI:15882
Certified
Videos
SDS
Documents

Documents