missing translation for 'onlineSavingsMsg'
Learn More

Phenol Red Sodium Salt, TCI America™

Catalog No. P01025G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P01025G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P01025G Supplier TCI America Supplier No. P01025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 34487-61-1
Molecular Formula C19H13NaO5S
Molecular Weight (g/mol) 376.358
MDL Number MFCD00066901
InChI Key HKHYOKBQJILTEI-UHFFFAOYSA-M
Synonym phenol red, sodium salt, phenolsulfonephthalein sodium salt, sodium 4-3-4-hydroxyphenyl-1,1-dioxido-3h-benzo c 1,2 oxathiol-3-yl phenolate, phenol red sodium salt, phenol, 4,4'-1,1-dioxido-3h-2,1-benzoxathiol-3-ylidene bis-, monosodium salt, phenol, 4,4'-1,1-dioxido-3h-2,1-benzoxathiol-3-ylidene bis-, sodium salt 1:1, phenol red, acs, phenolredsodiumsalt, phenolsulfonephthalein sodium, sodium 4-3-4-hydroxyphenyl-1,1-dioxo-2,1??-benzoxathiol-3-yl benzenolate
PubChem CID 23686673
IUPAC Name sodium;4-[3-(4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenolate
SMILES C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC=C(C=C3)O)C4=CC=C(C=C4)[O-].[Na+]

Specifications

Boiling Point 285°C
Quantity 5 g
Formula Weight 376.36
Physical Form Crystalline Powder
Chemical Name or Material Phenol Red Sodium Salt