missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenol Red Indicator, 0.04% (w/v), Ricca Chemical
Spécifications
Spécifications
| Chemical Name or Material | Phenol Red Indicator |
| CAS | 7732-18-5, 34487-61-1 |
| Percent Purity | 0.04% W/V |
| Grade | Indicator |
| Concentration or Composition (by Analyte or Components) | 0.04% (w/v) Aqueous Solution |
| Packaging | Natural Poly Bottle |
| Color | Orange/Red |
| pH | 6 to 7.3 |
| Quantity | 120 mL |