missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenol, Liquefied, Technical, 88%, Spectrum™ Chemical
$1058.47 - $15180.12
Chemical Identifiers
| CAS | 108-95-2 |
|---|---|
| Molecular Formula | C6H6O |
| Molecular Weight (g/mol) | 94.11 |
| InChI Key | ISWSIDIOOBJBQZ-UHFFFAOYSA-N |
| IUPAC Name | phenol |
| SMILES | OC1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18-612-259
|
Spectrum Chemical Mfg Cor
P10714LTPV |
4 L | Poly-Coated Glass Bottle |
Each for $1,058.47
|
|
||||
|
18-612-229
|
Spectrum Chemical Mfg Cor
P1071200LTBL |
200 L | Drum |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Specifications
| 108-95-2,7732-18-5 | |
| C6H6O | |
| ISWSIDIOOBJBQZ-UHFFFAOYSA-N | |
| phenol | |
| 0.88 |
| Poly-Coated Glass Bottle | |
| 4 L | |
| OC1=CC=CC=C1 | |
| 94.11 | |
| Technical |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.