missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenol, Liquefied, Technical, 88%, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor P1071200LTBL
Specifications
| 108-95-2,7732-18-5 | |
| C6H6O | |
| ISWSIDIOOBJBQZ-UHFFFAOYSA-N | |
| phenol | |
| 0.88 |
| Drum | |
| 200 L | |
| OC1=CC=CC=C1 | |
| 94.11 | |
| Technical |
Chemical Identifiers
| 108-95-2 | |
| 94.11 | |
| phenol |
| C6H6O | |
| ISWSIDIOOBJBQZ-UHFFFAOYSA-N | |
| OC1=CC=CC=C1 |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.