missing translation for 'onlineSavingsMsg'
Learn More

Phenol, Liquefied, Technical, 88%, Spectrum™ Chemical

Catalog No. 18612259
Change view
Click to view available options
Quantity:
4 L
200 L
Packaging:
Drum
Poly-Coated Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18612259 4 L Poly-Coated Glass Bottle
18612229 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18612259 Supplier Spectrum Chemical Mfg Cor Supplier No. P10714LTPV

Chemical Identifiers

CAS 108-95-2 , 7732-18-5
Molecular Formula C6H6O
Molecular Weight (g/mol) 94.11
InChI Key ISWSIDIOOBJBQZ-UHFFFAOYSA-N
IUPAC Name phenol
SMILES OC1=CC=CC=C1

Specifications

Packaging Poly-Coated Glass Bottle
Quantity 4 L
Percent Purity 0.88
Grade Technical

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.