missing translation for 'onlineSavingsMsg'
Learn More

Phenol, Liquefied, 89%, Spectrum™ Chemical

Catalog No. 18612258
Change view
Click to view available options
Quantity:
120 mL
500 mL
2.5 L
4 L
200 L
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18612258 4 L Amber Glass Bottle
18612224 120 mL Amber Glass Bottle
18612227 500 mL Amber Glass Bottle
18612225 2.5 L Amber Glass Bottle
18612226 200 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18612258 Supplier Spectrum Chemical Mfg Cor Supplier No. P10704LTPV
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 108-95-2 , 7732-18-5

Specifications

Packaging Amber Glass Bottle
Quantity 4 L
Residue after Evaporation 0.0005
Percent Purity 0.89
Grade Reagent
Physical Form Liquid

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.