missing translation for 'onlineSavingsMsg'
Learn More

Phenol, Liquefied, 89%, Spectrum™ Chemical

Catalog No. 18612227
Change view
Click to view available options
Quantity:
120 mL
500 mL
2.5 L
4 L
200 L
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18612227 500 mL Amber Glass Bottle
18612224 120 mL Amber Glass Bottle
18612225 2.5 L Amber Glass Bottle
18612258 4 L Amber Glass Bottle
18612226 200 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18612227 Supplier Spectrum Chemical Mfg Cor Supplier No. P1070500MLGL
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 108-95-2 , 7732-18-5

Specifications

Packaging Amber Glass Bottle
Quantity 500 mL
Residue after Evaporation 0.0005
Percent Purity 0.89
Grade Reagent
Physical Form Liquid

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.