missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenol/Chloroform/Isoamyl Alcohol (25:24:1 Mixture, pH 6.7/8.0, Liq.), Fisher BioReagents™
For extraction of proteins from crude nucleic acid preparations
Supplier: Fisher BioReagents BP1752I400
Description
For extraction of proteins from crude nucleic acid preparations
This mixture is used for extracting protein from crude nucleic acid preparations. Solution comes with a bottle of tris that should be used to equilibrate the pH.Greener Choice Claims
- Environmental benefits include:
- Recyclable
- Renewable Materials
Specifications
Phenol/Chloroform/Isoamyl Alcohol | |
108-95-2 | |
C6H6O | |
1.280g/cm³ | |
ISWSIDIOOBJBQZ-UHFFFAOYSA-N | |
phenol | |
996 | |
≥99.0% | |
Coated Amber Glass/Poison Pack | |
400 mL |
Clear | |
≥99.0 | |
MFCD00002143 | |
carbolic acid, hydroxybenzene, phenic acid, phenylic acid, oxybenzene, benzenol, phenyl hydrate, monophenol, phenyl hydroxide, phenylic alcohol | |
OC1=CC=CC=C1 | |
94.11 | |
CHEBI:15882 | |
Pass Test | |
6.8 to 8.2 | |
Liquid |
Chemical Identifiers
108-95-2 | |
94.11 | |
ISWSIDIOOBJBQZ-UHFFFAOYSA-N | |
996 | |
phenol |
C6H6O | |
MFCD00002143 | |
carbolic acid, hydroxybenzene, phenic acid, phenylic acid, oxybenzene, benzenol, phenyl hydrate, monophenol, phenyl hydroxide, phenylic alcoholShow More | |
CHEBI:15882 | |
OC1=CC=CC=C1 |
Safety and Handling

Emergency Overview
Toxic by inhalation, in contact with skin and if swallowed. Causes severe eye and skin irritation with possible burns. Irritating to respiratory system. Danger of serious damage to health by prol
Signal Word: DANGER!
DOTInformation : DOT Class 6.1, : Poison
Recommended Storage : 4°C