missing translation for 'onlineSavingsMsg'
Learn More

Phenol/Chloroform/Isoamyl Alcohol (25:24:1 Mixture, pH 6.7/8.0, Liq.), Fisher BioReagents™

Catalog No. p-27609
Change view
Click to view available options
Quantity:
100 mL
400 mL
Packaging:
Coated Amber Glass/Poison Pack
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP1752I100 100 mL Coated Amber Glass/Poison Pack
BP1752I400 400 mL Coated Amber Glass/Poison Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BP1752I100 Supplier Fisher BioReagents Supplier No. BP1752I 100
Only null left

For extraction of proteins from crude nucleic acid preparations

This mixture is used for extracting protein from crude nucleic acid preparations. Solution comes with a bottle of tris that should be used to equilibrate the pH.

Chemical Identifiers

CAS 108-95-2
Molecular Formula C6H6O
Molecular Weight (g/mol) 94.11
MDL Number MFCD00002143
InChI Key ISWSIDIOOBJBQZ-UHFFFAOYSA-N
Synonym carbolic acid, hydroxybenzene, phenic acid, phenylic acid, oxybenzene, benzenol, phenyl hydrate, monophenol, phenyl hydroxide, phenylic alcohol
PubChem CID 996
ChEBI CHEBI:15882
IUPAC Name phenol
SMILES OC1=CC=CC=C1

Specifications

Color Clear
Assay Percent Range ≥99.0
Density 1.280g/cm³
Percent Purity ≥99.0%
Identification Pass Test
Packaging Coated Amber Glass/Poison Pack
pH 6.8 to 8.2
Quantity 100 mL
Physical Form Liquid
Chemical Name or Material Phenol/Chloroform/Isoamyl Alcohol
Hazard Category Signal Word: DANGER!
Precautionary Statement Emergency Overview
Toxic by inhalation, in contact with skin and if swallowed. Causes severe eye and skin irritation with possible burns. Irritating to respiratory system. Danger of serious damage to health by prol
DOTInformation DOT Class 6.1, : Poison
Recommended Storage 4°C