missing translation for 'onlineSavingsMsg'
Learn More

Phenol, Certified, 5.0% (w/v) ±0.1% (w/v), LabChem™

Catalog No. p-5388326
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Amber Glass
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC181951 500 mL Amber Glass
LC181952 1 L Amber Glass
LC181954 4 L Amber Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. LC181951 Supplier LabChem Supplier No. LC181951
Only null left

Chemical Identifiers

CAS 108-95-2 , 7732-18-5
Molecular Formula C6H6O
Molecular Weight (g/mol) 94.11
MDL Number MFCD00002143
InChI Key ISWSIDIOOBJBQZ-UHFFFAOYSA-N
Synonym carbolic acid, hydroxybenzene, phenic acid, phenylic acid, oxybenzene, benzenol, phenyl hydrate, monophenol, phenyl hydroxide, phenylic alcohol
PubChem CID 996
ChEBI CHEBI:15882
IUPAC Name phenol
SMILES OC1=CC=CC=C1

Specifications

CAS Max % 95, 5
Decomposition Information Carbon monoxide; Carbon dioxide; Thermal decomposition generates: Corrosive vapors
Color Colorless
Density 1g/mL
Packaging Amber Glass
Linear Formula C6H5O8
Quantity 500 mL
Specific Gravity 1g/mL
Solubility Information Soluble in water
Formula Weight 94.11
Percent Purity 5.0% ±0.1% (w/v)
Grade Certified
Physical Form Liquid
Chemical Name or Material Phenol
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Suspected of causing genetic defects.
May cause damage to organs (liver, kidneys) through prolonged or repeated exposure.
Harmful to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Do not breathe mist, vapors, spray.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If exposed or concerned: Get medical advice/attention.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature