Learn More
Phenol, Certified, 5.0% (w/v) ±0.1% (w/v), LabChem™
Supplier: LabChem LC181952
Specifications
| Phenol | |
| 95,5 | |
| Colorless | |
| Amber Glass | |
| C6H5O8 | |
| 1 L | |
| carbolic acid, hydroxybenzene, phenic acid, phenylic acid, oxybenzene, benzenol, phenyl hydrate, monophenol, phenyl hydroxide, phenylic alcohol | |
| ISWSIDIOOBJBQZ-UHFFFAOYSA-N | |
| phenol | |
| 996 | |
| 94.11 | |
| Certified |
| 108-95-2,7732-18-5 | |
| Carbon monoxide; Carbon dioxide; Thermal decomposition generates: Corrosive vapors | |
| 1g/mL | |
| C6H6O | |
| MFCD00002143 | |
| 1g/mL | |
| Soluble in water | |
| OC1=CC=CC=C1 | |
| 94.11 | |
| CHEBI:15882 | |
| 5.0% ±0.1% (w/v) | |
| Liquid |
Chemical Identifiers
| 108-95-2 | |
| 94.11 | |
| ISWSIDIOOBJBQZ-UHFFFAOYSA-N | |
| 996 | |
| phenol |
| C6H6O | |
| MFCD00002143 | |
| carbolic acid, hydroxybenzene, phenic acid, phenylic acid, oxybenzene, benzenol, phenyl hydrate, monophenol, phenyl hydroxide, phenylic alcohol | |
| CHEBI:15882 | |
| OC1=CC=CC=C1 |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Suspected of causing genetic defects.
May cause damage to organs (liver, kidneys) through prolonged or repeated exposure.
Harmful to aquatic life with long lasting effects.
GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Do not breathe mist, vapors, spray.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If exposed or concerned: Get medical advice/attention.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Danger
Recommended Storage : Room Temperature