missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Pheneturide

Catalog No. AC381440050
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC381440050 5 g
AC381440250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC381440050 Supplier Thermo Scientific Chemicals Supplier No. 381440050
Only null left

CAS: 90-49-3 | C11H14N2O2 | 206.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 90-49-3
Molecular Formula C11H14N2O2
Molecular Weight (g/mol) 206.25
InChI Key AJOQSQHYDOFIOX-UHFFFAOYNA-N
IUPAC Name (2-phenylbutanoyl)urea
SMILES CCC(C(=O)NC(N)=O)C1=CC=CC=C1

Specifications

Melting Point 149.0°C to 152.0°C
Linear Formula NH2C(O)NHC(O)CH(C6H5)CH2CH3
Quantity 5 g
Merck Index 15, 7344
Solubility Information Solubility aq. soln.: < 0.5 g/L Other solubilities: soluble in other organic solvents
Formula Weight 206.24
Chemical Name or Material Pheneturide
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Harmful if swallowed.

Precautionary Statement GHS P Statement IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

Call a POISON CENTER or doctor/physician if you feel unwell.

Wash face, hands and any exposed skin thoroughly after handling.

EINECSNumber 201-998-2

RUO – Research Use Only