missing translation for 'onlineSavingsMsg'
Learn More

Phenethyl Isocyanate 98.0+%, TCI America™

Catalog No. P16775G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P16775G 5 g
P167725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P16775G Supplier TCI America Supplier No. P16775G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1943-82-4
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.177
MDL Number MFCD00037044
InChI Key HACRKYQRZABURO-UHFFFAOYSA-N
Synonym phenethyl isocyanate, 2-isocyanatoethyl benzene, phenylethyl isocyanate, 2-phenylethyl isocyanate, 2-phenylethylisocyanate, phenylethylisocyanate, phenethylisocyanate, benzene, 2-isocyanatoethyl, 2-isocyanato-ethyl-benzene, 2-phenethyl isocyanate
PubChem CID 160602
ChEBI CHEBI:60067
IUPAC Name 2-isocyanatoethylbenzene
SMILES C1=CC=C(C=C1)CCN=C=O

Specifications

Color Yellow
Boiling Point 210°C
Quantity 5 g
UN Number 2206
Formula Weight 147.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Phenethyl Isocyanate