missing translation for 'onlineSavingsMsg'
Learn More

Phenethyl Caffeate 98.0+%, TCI America™

Catalog No. P208825MG
Change view
Click to view available options
Quantity:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P208825MG 25 mg
P2088250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P208825MG Supplier TCI America Supplier No. P208825MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 104594-70-9
Molecular Formula C17H16O4
Molecular Weight (g/mol) 284.31
MDL Number MFCD00866470
InChI Key SWUARLUWKZWEBQ-VQHVLOKHSA-N
Synonym caffeic acid phenethyl ester, phenethyl caffeate, cape, capeee, phenethyl 3-3,4-dihydroxyphenyl acrylate, phenylethyl caffeate, unii-g960r9s5sk, caffeic acid phenylethyl ester, 2-phenylethyl caffeate, caffeic acid 2-phenylethyl ester
PubChem CID 5281787
ChEBI CHEBI:8062
IUPAC Name 2-phenylethyl (2E)-3-(3,4-dihydroxyphenyl)prop-2-enoate
SMILES OC1=CC=C(\C=C\C(=O)OCCC2=CC=CC=C2)C=C1O

Specifications

Melting Point 129°C
Color White-Yellow
Quantity 25 mg
Formula Weight 284.31
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Phenethyl Caffeate
RTECSNumber UD3334375
TSCA No
Recommended Storage Freezer