missing translation for 'onlineSavingsMsg'
Learn More

Phencyclone, 97%

Numéro de catalogue. AAL0010803
Modifier l'affichage
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
AAL0010803 1 g
AAL0010806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. AAL0010803 Fournisseur Thermo Scientific Chemicals Code fournisseur L0010803
Il en reste null

CAS: 5660-91-3 | C29H18O | 382.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Identifiants chimiques

CAS 5660-91-3
Molecular Formula C29H18O
Molecular Weight (g/mol) 382.46
MDL Number MFCD00051451
InChI Key MNSDGJFEKUKHGO-UHFFFAOYSA-N
Synonym phencyclone, 1,3-diphenyl-2h-cyclopenta l phenanthren-2-one, 1,3-diphenylcyclopenta l phenanthren-2-one, 1,3-diphenylcyclopenta 1,2-l phenanthren-2-one
PubChem CID 97140
ChEBI CHEBI:48304
IUPAC Name 1,3-diphenylcyclopenta[l]phenanthren-2-one
SMILES O=C1C(=C2C(=C1C1=CC=CC=C1)C1=CC=CC=C1C1=CC=CC=C21)C1=CC=CC=C1

Spécifications

Melting Point 245°C to 249°C
Quantity 1 g
Beilstein 2061817
Formula Weight 382.46
Percent Purity 97%
Chemical Name or Material Phencyclone
EINECSNumber 227-112-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only