missing translation for 'onlineSavingsMsg'
Learn More

Phenanthridine 98.0+%, TCI America™

Catalog No. P104710G
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P104710G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P104710G Supplier TCI America Supplier No. P104710G

Chemical Identifiers

CAS 229-87-8
Molecular Formula C13H9N
Molecular Weight (g/mol) 179.222
MDL Number MFCD00004989
InChI Key RDOWQLZANAYVLL-UHFFFAOYSA-N
Synonym 6-phenanthridine, benzo c quinoline, 3,4-benzoquinoline, 9-azaphenanthrene, 3,4-benzoisoquinoline, 5-azaphenanthrene, unii-62qgs7cps6, ccris 1234, 62qgs7cps6
PubChem CID 9189
ChEBI CHEBI:36421
IUPAC Name phenanthridine
SMILES C1=CC=C2C(=C1)C=NC3=CC=CC=C23

Specifications

Melting Point 108°C
Boiling Point 349°C
Quantity 10 g
Formula Weight 179.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Phenanthridine
RTECSNumber SF7928000
TSCA No