missing translation for 'onlineSavingsMsg'
Learn More

Phenanthrene Zone Refined (number of passes:30) 98.5+%, TCI America™

Catalog No. P0331ZONE
Change view
Click to view available options
Quantity:
ZONE
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0331ZONE ZONE
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P0331ZONE Supplier TCI America Supplier No. P03311SAMPLE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 85-01-8
Molecular Formula C14H10
Molecular Weight (g/mol) 178.23
MDL Number MFCD00001168
InChI Key YNPNZTXNASCQKK-UHFFFAOYSA-N
Synonym phenanthren, phenanthrin, phenanthracene, ravatite, phenantrin, phenanthren german, phenanthrene, pure, unii-448j8e5bst, ccris 1233, chembl46730
PubChem CID 995
ChEBI CHEBI:28851
IUPAC Name phenanthrene
SMILES C1=CC=C2C(C=CC3=CC=CC=C23)=C1

Specifications

Melting Point 100°C
Color White
Boiling Point 340°C
Quantity ZONE
UN Number 3077
Formula Weight 178.23
Percent Purity ≥98.5% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Phenanthrene Zone Refined (number of passes:30)
EINECSNumber (4)-0635
RTECSNumber SF7175000
TSCA Yes