missing translation for 'onlineSavingsMsg'
Learn More

Phenanthrene 97.0+%, TCI America™

Catalog No. P0079500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0079500G 500 g
P007925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P0079500G Supplier TCI America Supplier No. P0079500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 85-01-8
Molecular Formula C14H10
Molecular Weight (g/mol) 178.23
MDL Number MFCD00001168
InChI Key YNPNZTXNASCQKK-UHFFFAOYSA-N
Synonym phenanthren, phenanthrin, phenanthracene, ravatite, phenantrin, phenanthren german, phenanthrene, pure, unii-448j8e5bst, ccris 1233, chembl46730
PubChem CID 995
ChEBI CHEBI:28851
IUPAC Name phenanthrene
SMILES C1=CC=C2C(C=CC3=CC=CC=C23)=C1

Specifications

Melting Point 100°C
Boiling Point 340°C
Quantity 500 g
UN Number 3077
Formula Weight 178.23
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Phenanthrene
EINECSNumber (4)-0635
RTECSNumber SF7175000
TSCA Yes