missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenanthrene, 90%, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Boiling Point | 340°C/644°F |
| Packaging | Poly Bottle |
| Quantity | 500 g |
| Molecular Weight (g/mol) | 178.22 |
| Percent Purity | 0.9 |
| Grade | Reagent |
| Physical Form | Powder or Waxy Solid |
| Chemical Name or Material | Phenanthrene |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.