missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenacyl 4-(Bromomethyl)phenylacetate 97.0+%, TCI America™
Specifications
Specifications
| Melting Point | 88°C |
| Color | White-Yellow |
| Quantity | 5 g |
| Formula Weight | 347.21 |
| Percent Purity | ≥97.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Phenacyl 4-(Bromomethyl)phenylacetate |
Safety and Handling
| TSCA | No |