Learn More
Promega pHAb Amine Reactive Dye, 1 x 250microg
Catalog No. PRG9841
Encompass
Change view
Click to view available options
Quantity:
1 x 250 μg
Catalog No. Quantity
PRG9841 1 x 250 μg
1 options
Catalog No. PRG9841 Supplier Promega Supplier No. G9841
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
pHAb Amine Reactive Dye exhibits very low fluorescence at pH > 7 and much higher fluorescence as the pH of the solution becomes acidic. pHAb Dyes are designed specifically for antibody labeling.
  • Measure Antibody Internalization in Real Time
  • Compatible with 96−well, plate−based assay
  • High signal−to−background ratios
  • On−Bead conjugation
Specifications
Quantity 1 x 250 μg
Content And Storage <-65°C