missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ PHA 568487
Description
Agonist of the α7 nicotinic acetylcholine receptor (Ki values are 44 and 2800 for α7 and 5-HT3 respectively, IC50 values are > 100 μM for α3β4 and α1β1δγ respectively, and % inhibition is < 1 and 5% for α4β2 and hERG respectively). Orally active and brain penetrant.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 404.41 |
| Purity | >99% |
| Molecular Weight (g/mol) | 404.419 |
| Chemical Name or Material | PHA 568487 |
Safety and Handling
| Recommended Storage | Desiccate at Room Temperature |